missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HSD17B12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553418
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553418 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553418 Supplier Invitrogen™ Supplier No. PA553418
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: WGVGNEAGVG PGLGEWAVVT GSTDGIGKSY AEELAKHGMK VVLISRSKDK LDQVSSEIKE KFKVETRTIA VDFASEDIYD KIKTGLAGLE IGILVNNVGM SYEYPEYFLD VPDLD Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.

The enzyme 17-beta hydroxysteroid dehydrogenase-12 (HSD17B12) uses NADPH to reduce 3-ketoacyl-CoA to 3-hydroxyacyl-CoA during the second step of fatty acid elongation.

Specifications

Antigen HSD17B12
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HSD17B12
Gene Accession No. Q53GQ0
Gene Alias 17-beta-HSD 12; 17beta-HSD type 12; 17-beta-hydroxysteroid dehydrogenase 12; 2610510O05Rik; 3-ketoacyl-CoA reductase; AI172963; Estradiol 17-beta-dehydrogenase 12; HSD17B12; hydroxysteroid (17-beta) dehydrogenase 12; hydroxysteroid 17-beta dehydrogenase 12; KAR; keratoadhesin; keratonectin; Kik1; KIK-I; SDR12C1; short chain dehydrogenase/reductase family 12C member 1; short chain dehydrogenase/reductase family 12C, member 1; smooth muscle-specific 17 beta-hydroxysteroid dehydrogenase type 3; steroid dehydrogenase; steroid dehydrogenase homolog; very-long-chain 3-oxoacyl-CoA reductase
Gene Symbols HSD17B12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HSD17B12. Recombinant protein control fragment (Product #RP-91081).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51144
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less