missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSC70 Monoclonal Antibody (N27F3-4)

Description
MA1-10890 detects Heat Shock Protein 70/Heat Shock Cognate Protein 70 in human, mouse, rat, rabbit, monkey, guinea pig, hamster, chicken, bovine, canine, ovine, porcine, fish, Xenopus, Drosophila, C. elegans, and plant samples. MA1-10890 has been successfully used in Western blot, immunohistochemistry, immunoprecipitation and flow cytometry procedures. By Western blot, MA1-10890 is specific for the ~72 kDa Heat Shock Protein 70/Heat Shock Cognate Protein 70 protein. Positive control of HeLa cell lysate recommended. MA1-10890 immunogen is purified recombinant Hsp70/Hsc70.
The HSP70 family is a set of highly conserved proteins that are induced by a variety of biological stresses, including heat stress, in every organism in which the proteins have been examined. The human HSP70 family members include: HSP70, a protein which is strongly inducible in all organisms but which is also constitutively expressed in primate cells; HSP72, a 72 kDa protein that is induced exclusively under stress conditions; HSC70, or cognate protein, is a 72 kDa, constitutively expressed, protein which is involved in the uncoating of clathrin coated vesicles; GRP78, or BiP, is a glucose regulated 78 kDa protein localized in the endoplasmic reticulum; and p75, or HSP75, a 75 kDa protein that is found within the mitochondria. HSC70 (also known as HSC71, HSC73, HSP73, p72, prp73) is expressed constitutively and is slightly heat-inducible. HSC70 binds to the exposed loop of clathrin light chains to promote uncoating and can also bind the cytoskeleton which may facilitate cytoskeletal rearrangements. HSC70 has been shown to stimulate lysosomal degradation of intracellular proteins and to retard both aggregation and folding of mitochondrial precursor proteins in vitro.
Specifications
Specifications
| Antigen | HSC70 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | N27F3-4 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.4 |
| Gene | Hspa8 |
| Gene Accession No. | P11142, P63017, P63018 |
| Gene Alias | 2410008N15Rik; AAG2; constitutive heat shock protein 70; constitutively expressed heat shock protein; epididymis luminal protein 33; epididymis secretory sperm binding protein Li 72p; FAM10A1; Heat shock 70 kDa protein 8; heat shock 70kd protein 10; heat shock 70kd protein 10 (HSC71); heat shock 70kD protein 8; heat shock 70kDa protein 8; heat shock cognate; heat shock cognate 70; heat shock cognate 71 kD protein; heat shock cognate 71 kDa protein; heat shock cognate 71 kDa protein-like protein; heat shock cognate 71-kd protein; heat shock cognate hsc73; heat shock cognate protein 54; Heat shock cognate protein 70; heat shock protein 70 cognate; heat shock protein 8; heat shock protein A8; heat shock protein cognate 70; heat shock protein family A (Hsp70) member 8; HEL-33; HEL-S-72p; HIP; HSC54; Hsc70; Hsc70 ATPase; HSC70 protein; Hsc71; Hsc73; HSP70; HSP71; HSP73; Hspa10; Hspa8; hspa8 protein; I79_022974; LAP; LAP1; LAP-1; lipopolysaccharide-associated protein 1; LPS-associated protein 1; MGC131511; MGC29929; NIP71; N-myristoyltransferase inhibitor protein 71; Protein FAM10A1; similar to 7O KDa heat shock cognate protein; SNC6; unnamed protein product; wu:fb01g06; wu:fi48b06 |
| Show More |