Learn More
Applied Biosystems™ HRM Software Module for QuantStudio™ 12K Flex System
Catalog No. 4472054
Encompass
Change view
Click to view available options
Quantity:
1 kit
Catalog No. Quantity
4472054 1 kit
1 options
Catalog No. 4472054 Supplier Applied Biosystems™ Supplier No. 4472054
This software module expands the functionality of the QuantStudio™ 12K Flex software, allowing high resolution melting (HRM) analysis to be performed on PCR amplicons.
This software module expands the functionality of the QuantStudio™ 12K Flex software, allowing high resolution melting (HRM) analysis to be performed on PCR amplicons.
High Resolution Melt (HRM) analysis is a new, post-PCR analysis method used for identifying genetic variation in nucleic acid sequences. Simple and fast, this method is based on PCR melt (dissociation) curve techniques and is enabled by the recent availability of improved double-stranded DNA (dsDNA)-binding dyes along with next-generation real-time PCR instrumentation and analysis software. HRM analysis can discriminate DNA sequences based on their composition, length, GC content, or strand complementarity.
Specifications
For Use With (Equipment) QuantStudio™ 12 k Flex
Product Line QuantStudio
Operating System Windows 7
License 10 Seat
Quantity 1 kit
What is the difference between continuous melt curves and step and hold melt curves on the Applied Biosystems Real-Time PCR Instruments?
Continuous melt curve: increases the temperature by the ramp increment (degrees C/sec) so that the temperature will change at a constant rate set in the protocol.
Step and Hold melt curve: increases the temperature by the ramp increment (degrees C) and then holds at that temperature for the time specified by the user.
The amount of data collected will depend on the filter(s) selected. The more filters you use, the longer the acquisition will take and the fewer data points will be collected.
What type of Real-Time PCR (Sequence Detection System) information is available from Applied Biosystems?
Tutorials demonstrating introductions to Real-Time PCR, guidelines for designing real-time assays using Primer Express software, and various instrument set-ups can be found under Life Technologies University on our website. User bulletins, manuals and Product inserts, protocols can be found as downloadable pdf files on our Technical Resources page.
For Research Use Only. Not for use in diagnostic procedures.