missing translation for 'onlineSavingsMsg'
Learn More

Acetone, HR-GC, 99.7% min, OmniSolv, MilliporeSigma™

Catalog No. MAX01101
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAX01101 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAX01101 Supplier Technidata Supplier No. AX01101

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Quantity 4 L
Physical Form Liquid
Assay Percent Range 99.7%, min.
Linear Formula CH3COCH3
Formula Weight 58.08
Chemical Name or Material HR-GC Acetone, HR-GC, OmniSolv™