missing translation for 'onlineSavingsMsg'
Learn More

HPLC Water, MilliporeSigma™

Catalog No. MWX00081
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MWX00081 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MWX00081 Supplier Technidata Supplier No. WX00081

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Absorbance At 300-400nm: 0.005AU max
Color Colorless
Boiling Point 100°C
Linear Formula H2O
Formula Weight 18.2
Grade HPLC
Residue after Evaporation 2ppm max
Vapor Pressure 23 hPa (20°C)
Quantity 4 L
Refractive Index 1.3320 to 1.3340
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
RTECSNumber ZC0110000
Recommended Storage Room Temperature