missing translation for 'onlineSavingsMsg'
Learn More

Restek™ HPLC Stainless Steel Capillary Tubing Assemblies

For Agilent HPLC Systems.

Supplier:  Restek™ RK 26534

Encompass

 View more versions of this product

Catalog No. 06728959


Only null left

Description

Description

  • Precut, micropolished tubing and preseated fittings for quick, easy maintenance of your Agilent HPLC systems
  • Material: Stainless steel
Specifications

Chemical Identifiers

Agilent HPLC Systems

Specifications

Agilent HPLC Systems
PreSeated fittings for quick, easy maintenance
Smooth
0.006 in.
150 mm
Capillary Tubing
1100
HPLC Systems
5.9 in.
Stainless Steel (316)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions