missing translation for 'onlineSavingsMsg'
Learn More

Restek™ HPLC Stainless Steel Capillary Tubing Assemblies

For Agilent HPLC Systems.

Supplier:  Restek™ RK 26528

Encompass

 View more versions of this product

Catalog No. 06728953


Only null left

Description

Description

  • Precut, micropolished tubing and preseated fittings for quick, easy maintenance of your Agilent HPLC systems
  • Material: Stainless steel
Specifications

Chemical Identifiers

Agilent HPLC Systems

Specifications

Agilent HPLC Systems
Blue
1050
2.2 ft.
Stainless Steel
Capillary Tubing with Fitting
Preseated fittings 0.06 in. Stainless Steel Male Nut, F/B Ferrule Set
0.009 in.
700 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions