missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HPCAL1 Monoclonal Antibody (K52064_13F1), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA556000
Description
Sequence of this protein is as follows: MGKQNSKLRP EVLQDLRENT EFTDHELQEW YKGFLKDCPT GHLTVDEFKK IYANFFPYGD ASKFAEHVFR TFDTNGDGTI DFREFIIALS VTSRGKLEQK LKWAFSMYDL DGNGYISRSE MLEIVQAIYK MVSSVMKMPE DESTPEKRTD KIFRQMDTNN DGKLSLEEFI RGAKSDPSIV RLLQCDPSSA SQF
Hippocalcin-like protein 1, may be involved in the calcium-dependent regulation of rhodopsin phosphorylation.Specifications
| HPCAL1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P37235, P62748, P62749 | |
| HPCAL1 | |
| Recombinant human HPCAL1 (immunogen range is 1-193). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| K52064_13F1 | |
| Unconjugated | |
| HPCAL1 | |
| BDR1; Calcium-binding protein BDR-1; hippocalcin like 1; hippocalcin-like 1; hippocalcin-like 1; visinin like 3; neural visinin-like 3; neural visinin-like protein 3; hippocalcin-like protein 1; HLP2; Hpcal1; neural visinin-like 3; neural visinin-like Ca2+-binding protein type 3; neural visinin-like protein 3; NVL-3; Nvp3; NVP-3; VILIP3; VILIP-3; visinin like 3; visinin-like protein 3; Vnsl3; Vsnl3 | |
| Mouse | |
| Protein A | |
| RUO | |
| 3241, 50871, 53602 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |