missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HPCAL1 Monoclonal Antibody (K52064_13F1), Invitrogen™
Description
Sequence of this protein is as follows: MGKQNSKLRP EVLQDLRENT EFTDHELQEW YKGFLKDCPT GHLTVDEFKK IYANFFPYGD ASKFAEHVFR TFDTNGDGTI DFREFIIALS VTSRGKLEQK LKWAFSMYDL DGNGYISRSE MLEIVQAIYK MVSSVMKMPE DESTPEKRTD KIFRQMDTNN DGKLSLEEFI RGAKSDPSIV RLLQCDPSSA SQF
Hippocalcin-like protein 1, may be involved in the calcium-dependent regulation of rhodopsin phosphorylation.
Specifications
Specifications
| Antigen | HPCAL1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | K52064_13F1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide |
| Gene | HPCAL1 |
| Gene Accession No. | P37235, P62748, P62749 |
| Gene Alias | BDR1; Calcium-binding protein BDR-1; hippocalcin like 1; hippocalcin-like 1; hippocalcin-like 1; visinin like 3; neural visinin-like 3; neural visinin-like protein 3; hippocalcin-like protein 1; HLP2; Hpcal1; neural visinin-like 3; neural visinin-like Ca2+-binding protein type 3; neural visinin-like protein 3; NVL-3; Nvp3; NVP-3; VILIP3; VILIP-3; visinin like 3; visinin-like protein 3; Vnsl3; Vsnl3 |
| Show More |