missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HOXB9 Recombinant Rabbit Monoclonal Antibody (23GB6075)
GREENER_CHOICE

Catalog No. MA552980
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552980 100 μL
MA552981 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552980 Supplier Invitrogen™ Supplier No. MA552980

Rabbit Recombinant Monoclonal Antibody

HOXB9 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

HOXB9 belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. HOXB9 is one of several homeobox HOXB genes located in a cluster on chromosome 17. The exact role of this gene has yet to be determined.

Specifications

Antigen HOXB9
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB6075
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HOXB9
Gene Accession No. P17482
Gene Alias HGNC:5120; homeo box 2E; homeo box B9; homeobox B9; homeobox protein Hox-2.5; Homeobox protein Hox-2E; homeobox protein Hox-B9; HOX2; HOX-2.5; HOX2E; HOXB9; Hoxb-9; MGC124051; MGC124052; MGC124053; MGC124054
Gene Symbols HOXB9
Host Species Rabbit
Immunogen A synthesized peptide derived from human HOXB9 (1-52AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3219
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less