missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HOXB4 Recombinant Rabbit Monoclonal Antibody (23GB6620)

Catalog No. MA553079
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553079 20 μL
MA553078 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553079 Supplier Invitrogen™ Supplier No. MA553079
Only null left

Rabbit Recombinant Monoclonal Antibody

HOXB4 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

HOX homeobox genes play a role both in the early stem cell function as well as in later stages of hematopoietic differentiation. HOXB4, specifically, is a homeotic selector gene implicated in self-renewal of definitive HSCs. Expression of HOXB4 in primitive progenitors combined with culture on hematopoietic stroma induced a switch to the definitive HSC phenotype. Activation of WNT signaling in HSCs induced increased expression of HOXB4. Perturbations of HOX gene expression can cause leukemia.

Specifications

Antigen HOXB4
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB6620
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HOXB4
Gene Accession No. P17483
Gene Alias homeo box 2F; homeo box B4; homeo box B4a; homeobox B4; homeobox B4a; homeobox gene B-4; homeobox protein Hox-2.6; Homeobox protein Hox-2F; homeobox protein Hox-B4; Homeobox protein Hox-B4a; homeobox protein Zf-13; HOX2; Hox-2.6; HOX2F; Hoxb4; HOX-B4; Hoxb-4; Hoxb4 protein; hoxb4a; Hoxb4a variant 2; RGD1560113; sb:eu512; Z-17; zf13; ZF-13
Gene Symbols HOXB4
Host Species Rabbit
Immunogen A synthesized peptide derived from human HOXB4 (21-55AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3214, 497988
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less