missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HOXA5 Recombinant Rabbit Monoclonal Antibody (23GB6380)
GREENER_CHOICE

Catalog No. MA553032
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553032 100 μL
MA553033 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553032 Supplier Invitrogen™ Supplier No. MA553032
Only null left

Rabbit Recombinant Monoclonal Antibody

HOXA5 Recombinant Monoclonal Antibody for Western Blot

In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. HOXA5 gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. Methylation of this gene may result in the loss of its expression and, since the encoded protein upregulates the tumor suppressor p53, this protein may play an important role in tumorigenesis.

Specifications

Antigen HOXA5
Applications Western Blot
Classification Recombinant Monoclonal
Clone 23GB6380
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HOXA5
Gene Accession No. P20719
Gene Alias homeo box 1C; homeo box A5; homeobox A5; homeobox protein Hox-1.3; Homeobox protein Hox-1C; homeobox protein HOXA5; homeobox protein Hox-A5; homeobox protein M2; HOX1; HOX1.3; Hox-1.3; HOX1C; Hox-1C; Hoxa5; Hoxa-5; Unknown (protein for MGC:138007)
Gene Symbols HOXA5
Host Species Rabbit
Immunogen A synthesized peptide derived from human HOXA5 (100-150AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3202
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less