missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HOXA11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115376
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115376 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115376 Supplier Invitrogen™ Supplier No. PA5115376
Only null left

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HOXA11.

HOX11, also called homeobox11 or Tcl-3 is located at the 10q24 T-Cell translocation breakpoint region. It encodes a homeobox-domain containing protein, HOX11, containing a glycine and proline-rich amino terminus. HOX11 is required for maintenance of the developing spleen and cell survival. The HOX11protein binds to a specific DNA sequence and localizes to the cell nucleus. It transactivates transcription of a reporter gene linked to a cis-regulatory element, acting as a positive transcription activator. The HOX11 gene is highly expressed in T-ALLs as a result of a t (7:10) (q35;q24) or t (10:14) (q24;qll) translocation. The homeobox gene is deregulated upon translocation into the T-cell receptor locus and activates a specific subset of tumor-associated target genes. HOX 11 gene translocation has been attributed to T-cell leukemia and lymphoid neoplasias.

Specifications

Antigen HOXA11
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene HOXA11
Gene Accession No. P31270, P31311
Gene Alias homeo box 1I; homeo box A11; homeobox A11; homeobox protein Hox-1.9; Homeobox protein Hox-1I; homeobox protein HOXA11; homeobox protein Hox-A11; HOX1; Hox-1.9; HOX11; HOX1I; Hoxa11; Hoxa-11; NEWGENE_1564605; RUSAT1
Gene Symbols HOXA11
Host Species Rabbit
Immunogen A synthesized peptide derived from human HOXA11(Accession P31270), corresponding to amino acid residues P137-P187.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103692131, 15396, 3207
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less