missing translation for 'onlineSavingsMsg'
Learn More

SP Scientific Hotpack Glassware Washer Spindle Racks

Useful for cleaning inside narrow-neck glassware, these spindle racks are designed for use with undercounter, freestanding and Vertical Stacking SP Hotpack™ Glassware Washer models.

Supplier:  SP Scientific 446493

 View more versions of this product

Catalog No. 15365223

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • Recommended for graduated cylinders, volumetric flasks, round bottom flasks, Erlenmeyer flasks and other narrow-neck glassware
  • Multiple configurations available to suit a variety of glassware

NOTE: Spindle racks fit only in units that are spindle rack ready

Specifications

Chemical Identifiers

Stainless Steel
Washing Lab Glassware
24 in.
4 in.
24 in.
24 in.
Stainless Steel
Hotpack Spindle Rack Ready Washers Only
61 cm
10 cm
61 cm
61 cm

Specifications

Lower spindle rack mixed use
Lower Spindle Rack only
12 pins
Full Bottom Rack: 1/2 open and 1/2 with 12 spindles
Hotpack Spindle Rack Ready Washers Only
61 cm
24 in. x 24 in. x 4 in.
10 cm
61 cm
61 cm
2.2 kg
Stainless Steel
Stainless Steel
Replaces Bottom Open Rack; Must be used without top rack for tall glassware
Washing Lab Glassware
24 in.
24 in. x 24 in.
4 in.
24 in.
24 in.
5 lb.
Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions