missing translation for 'onlineSavingsMsg'
Learn More

Hordenine 98.0+%, TCI America™

Catalog No. H13515G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13515G 5 g
H13511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13515G Supplier TCI America Supplier No. H13515G

Chemical Identifiers

CAS 539-15-1
Molecular Formula C10H17NO5S
Molecular Weight (g/mol) 263.31
MDL Number MFCD00051462
InChI Key OIIQUBZPQJNHQK-UHFFFAOYSA-N
Synonym Anhaline, 4-[2-(Dimethylamino)ethyl]phenol, 4-Hydroxy-N,N-dimethylphenethylamine
PubChem CID 68313
ChEBI CHEBI:5764
IUPAC Name 4-[2-(dimethylamino)ethyl]phenol; sulfuric acid
SMILES OS(O)(=O)=O.CN(C)CCC1=CC=C(O)C=C1

Specifications

Melting Point 118°C
Color Yellow
Boiling Point 174°C
Quantity 5 g
Formula Weight 165.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Hordenine