CAS RN 539-15-1
CAS RN 539-15-1
IUPAC Name:
4-[2-(dimethylamino)ethyl]phenol; sulfuric acid
Synonyms:
hordenine
Molecular Weight (g/mol):
263.31
Molecular Formula:
C10H17NO5S
InChi Key:
OIIQUBZPQJNHQK-UHFFFAOYSA-N
SMILES:
OS(O)(=O)=O.CN(C)CCC1=CC=C(O)C=C1
1
–
1
of
1
results
1
Hordenine 98.0+%, TCI America™
CAS: 539-15-1 Molecular Formula: C10H17NO5S Molecular Weight (g/mol): 263.31 MDL Number: MFCD00051462 InChI Key: OIIQUBZPQJNHQK-UHFFFAOYSA-N Synonym: Anhaline, 4-[2-(Dimethylamino)ethyl]phenol, 4-Hydroxy-N,N-dimethylphenethylamine PubChem CID: 68313 ChEBI: CHEBI:5764 IUPAC Name: 4-[2-(dimethylamino)ethyl]phenol; sulfuric acid SMILES: OS(O)(=O)=O.CN(C)CCC1=CC=C(O)C=C1