Learn More
Eisco™ Hooked Cylinder
Hooked Cylinder for use with physics experiments.
Supplier:  Eisco™ PH0112D
 View more versions of this product
Catalog No. S12920
Only null left
Description
Description
  • Can be used for specific heat, specific gravity or density.
  • Used to study concepts related to physics.
Specifications
Chemical Identifiers
7 to 12
For use with physics experiments
1 Cylinder
Hooked Cylinder
5 cm x 13 mm
Used for specific heat, specific gravity, density
1 Lab Group
Specifications
7 to 12
For use with physics experiments
1 Cylinder
1 Lab Group
5 cm x 13 mm
Used for specific heat, specific gravity, density
Brass
Hooked Cylinder
Product Suggestions
Product Suggestions