missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Safety Products™ Saline Wall Station
Description
The Honewell Saline Wall Station is easily mounted in hazardous areas to provide immediate relief in case of eye injury.
Choose from 16 or 32 oz. (473 or 946 mL) bottles.
- Highly visible, with full emergency-use instructions on front
- Made from sturdy, high-impact plastic
- Bottles are single-use only with snap-off tops for quick and easy access
- Bottles have extended flow nozzles; 32 oz. bottles have integral grip for ease of use in an emergency
Order Info
Refill bottles sold separately. See Cat. Nos. 19-003-030 and -033.
Compliance
Does not meet the criteria of plumbed or self-contained eyewash equipment.
Specifications
Specifications
| Capacity (English) | 32 oz. |
| Capacity (Metric) | 946 mL |
| Certifications/Compliance | FDA |
| For Use With (Application) | Flushing Nuisance Particles Such as Pollen, Dust, Saw Dust, Smoke and More |
| For Use With (Equipment) | For Refill Bottles, See 19-003-030. |
| Includes | Sperian Saline Buffered, Saline solution superior to tap Water |
| Material | Polyethylene |
| Product Type | Double Eye Wall Station |