missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Fendall Flash Flood Eyewash Stations
Description
- Lightweight flash flood eyewash station delivers three-minute emergency eyewash
- Portable gravity-fed unit requires no plumbing or expensive installation
- Made of tough high-impact polyethylene
- Its bright green color is highly visible - even to those with impaired vision
- With just a pull of a strap, soothing Eyesaline™ solution provides immediate relief
- Additional Eyesaline refill cartridges available separately
Specifications
Specifications
| Capacity (English) | 128 oz. |
| Capacity (Metric) | 3785.41 mL |
| Depth (English) | 14 in. |
| Depth (Metric) | 35.56 cm |
| Height (English) | 20 in. |
| Height (Metric) | 5.08 cm |
| Includes | Buffered, pH-balanced Saline Cartridge |
| Product Type | Eyewash Station |