missing translation for 'onlineSavingsMsg'
Learn More

Homovanillic acid, 98%

CAS: 306-08-1 | C9H10O4 | 182.18 g/mol

$145.60 - $145.60

Chemical Identifiers

CAS 306-08-1
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00004350
InChI Key QRMZSPFSDQBLIX-UHFFFAOYSA-N
Synonym homovanillic acid, 4-hydroxy-3-methoxyphenylacetic acid, 2-4-hydroxy-3-methoxyphenyl acetic acid, vanillacetic acid, benzeneacetic acid, 4-hydroxy-3-methoxy, 3-methoxy-4-hydroxyphenylacetic acid, homovaniuic acid, 4-hydroxy-3-methoxyphenyl acetic acid, homovanillate, 4-hydroxy-3-methoxybenzeneacetic acid
PubChem CID 1738
ChEBI CHEBI:545959
IUPAC Name 2-(4-hydroxy-3-methoxyphenyl)acetic acid
SMILES COC1=C(C=CC(=C1)CC(=O)O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC162530010
SDS View Documents
Thermo Scientific Chemicals
162530010
1 g Glass bottle
Each for $145.60
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

306-08-1
182.18
QRMZSPFSDQBLIX-UHFFFAOYSA-N
1738
2-(4-hydroxy-3-methoxyphenyl)acetic acid
C9H10O4
MFCD00004350
homovanillic acid, 4-hydroxy-3-methoxyphenylacetic acid, 2-4-hydroxy-3-methoxyphenyl acetic acid, vanillacetic acid, benzeneacetic acid, 4-hydroxy-3-methoxy, 3-methoxy-4-hydroxyphenylacetic acid, homovaniuic acid, 4-hydroxy-3-methoxyphenyl acetic acid, homovanillate, 4-hydroxy-3-methoxybenzeneacetic acid
CHEBI:545959
COC1=C(C=CC(=C1)CC(=O)O)O

Specifications

306-08-1
White to Beige
97.5% min. (HPLC)
C9H10O4
MFCD00004350
10, 409
homovanillic acid, 4-hydroxy-3-methoxyphenylacetic acid, 2-4-hydroxy-3-methoxyphenyl acetic acid, vanillacetic acid, benzeneacetic acid, 4-hydroxy-3-methoxy, 3-methoxy-4-hydroxyphenylacetic acid, homovaniuic acid, 4-hydroxy-3-methoxyphenyl acetic acid, homovanillate, 4-hydroxy-3-methoxybenzeneacetic acid
QRMZSPFSDQBLIX-UHFFFAOYSA-N
2-(4-hydroxy-3-methoxyphenyl)acetic acid
1738
182.18
Crystalline Powder
141°C to 145°C
Authentic
Glass bottle
HOC6H3(OCH3)CH2CO2H
1 g
15, 4779
Solubility in water: soluble. Other solubilities: slightly soluble in alcohol,ether,soluble in benzene,insoluble in cyclohexane
COC1=C(C=CC(=C1)CC(=O)O)O
182.18
CHEBI:545959
98%
Homovanillic acid, 98%
Videos
Safety and Handling

Safety and Handling

AIR SENSITIVE

EINECSNumber : 206-176-7

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only