missing translation for 'onlineSavingsMsg'
Learn More

Homovanillic acid, 98%

Catalog No. AC162530010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC162530010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC162530010 Supplier Thermo Scientific Chemicals Supplier No. 162530010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 306-08-1 | C9H10O4 | 182.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 306-08-1
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00004350
InChI Key QRMZSPFSDQBLIX-UHFFFAOYSA-N
Synonym homovanillic acid, 4-hydroxy-3-methoxyphenylacetic acid, 2-4-hydroxy-3-methoxyphenyl acetic acid, vanillacetic acid, benzeneacetic acid, 4-hydroxy-3-methoxy, 3-methoxy-4-hydroxyphenylacetic acid, homovaniuic acid, 4-hydroxy-3-methoxyphenyl acetic acid, homovanillate, 4-hydroxy-3-methoxybenzeneacetic acid
PubChem CID 1738
ChEBI CHEBI:545959
IUPAC Name 2-(4-hydroxy-3-methoxyphenyl)acetic acid
SMILES COC1=C(C=CC(=C1)CC(=O)O)O

Specifications

Melting Point 141°C to 145°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula HOC6H3(OCH3)CH2CO2H
Quantity 1 g
Beilstein 10, 409
Merck Index 15, 4779
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in alcohol,ether,soluble in benzene,insoluble in cyclohexane
Formula Weight 182.18
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Homovanillic acid, 98%
Show More Show Less
Hazard Category AIR SENSITIVE
EINECSNumber 206-176-7
TSCA TSCA

RUO – Research Use Only