missing translation for 'onlineSavingsMsg'
Learn More

Homovanillic acid, 98%

Catalog No. AAA1244106
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1244106 5 g
AAA1244103 1 g
AAA1244114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1244106 Supplier Thermo Scientific Chemicals Supplier No. A1244106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 306-08-1 | C9H10O4 | 182.175 g/mol

Fluorimetric reagent and major catecholamine metaboliteHomovanillic acid is used as a reagent in the determination of oxidative enzymes. It is associated with dopamine levels in the brain. Its presence supports a diagnosis of neuroblastoma and malignant pheochromocytoma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water,dimethyl sulfoxide, ethanol, methanol and ethyl acetate.

Notes
Light sensitive. Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 306-08-1
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.175
MDL Number MFCD00004350
InChI Key QRMZSPFSDQBLIX-UHFFFAOYSA-N
Synonym homovanillic acid, 4-hydroxy-3-methoxyphenylacetic acid, 2-4-hydroxy-3-methoxyphenyl acetic acid, vanillacetic acid, benzeneacetic acid, 4-hydroxy-3-methoxy, 3-methoxy-4-hydroxyphenylacetic acid, homovaniuic acid, 4-hydroxy-3-methoxyphenyl acetic acid, homovanillate, 4-hydroxy-3-methoxybenzeneacetic acid
PubChem CID 1738
ChEBI CHEBI:545959
IUPAC Name 2-(4-hydroxy-3-methoxyphenyl)acetic acid
SMILES COC1=C(C=CC(=C1)CC(=O)O)O

Specifications

Melting Point 140°C to 145°C
Quantity 5 g
Beilstein 2213447
Merck Index 14,4740
Solubility Information Soluble in water,dimethyl sulfoxide,ethanol,methanol and ethyl acetate.
Formula Weight 182.18
Percent Purity ≥98%
Chemical Name or Material Homovanillic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 206-176-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only