missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Homosalate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Homosalate is a UV filter that protects the skin from the harmful effects of UVB (280-315 nm) and UVA II (315-340 nm) radiations. It is widely utilized in make-up, sunscreen and topical formulations.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 118-56-9 |
| Flash Point | 169°C to 173°C |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available. |
| Physical Form | Neat |
| Linear Formula | C16H22O3 |
| MDL Number | MFCD00019377 |
| Synonym | 3,3,5-Trimethylcyclohexyl salicylate; 2-Hydroxybenzoic acid 3,3,5-trimethylcyclohexyl ester; Homomenthyl salicylate; Homosalate |
| Type | Certified Reference Material |
| Quantity | 1 g |
| Formula Weight | 262.34 |
| Show More |