Learn More
Homophthalic acid, 99%
CAS: 89-51-0 | C9H8O4 | 180.159 g/mol
$125.70 - $430.74
Chemical Identifiers
| CAS | 89-51-0 |
|---|---|
| Molecular Formula | C9H8O4 |
| Molecular Weight (g/mol) | 180.159 |
| MDL Number | MFCD00004326 |
| InChI Key | ZHQLTKAVLJKSKR-UHFFFAOYSA-N |
| Synonym | homophthalic acid, 2-carboxymethyl benzoic acid, benzeneacetic acid, 2-carboxy, 2-carboxyphenylacetic acid, 2-carboxybenzeneacetic acid, alpha-carboxy-o-toluic acid, o-carboxymethyl benzoic acid, 2-carboxyphenyl acetic acid, o-toluic acid, alpha-carboxy, toluene-alpha,2-dicarboxylic acid |
| PubChem CID | 66643 |
| IUPAC Name | 2-(carboxymethyl)benzoic acid |
| SMILES | C1=CC=C(C(=C1)CC(=O)O)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1429718
|
Thermo Scientific Chemicals
A1429718 |
50 g |
Each for $125.70
|
|
|||||
|
AAA1429730
|
Thermo Scientific Chemicals
A1429730 |
250 g |
Each for $430.74
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 89-51-0 | |
| 180.159 | |
| ZHQLTKAVLJKSKR-UHFFFAOYSA-N | |
| 66643 | |
| C1=CC=C(C(=C1)CC(=O)O)C(=O)O |
| C9H8O4 | |
| MFCD00004326 | |
| homophthalic acid, 2-carboxymethyl benzoic acid, benzeneacetic acid, 2-carboxy, 2-carboxyphenylacetic acid, 2-carboxybenzeneacetic acid, alpha-carboxy-o-toluic acid, o-carboxymethyl benzoic acid, 2-carboxyphenyl acetic acid, o-toluic acid, alpha-carboxy, toluene-alpha,2-dicarboxylic acid | |
| 2-(carboxymethyl)benzoic acid |
Specifications
| 89-51-0 | |
| C9H8O4 | |
| 50 g | |
| homophthalic acid, 2-carboxymethyl benzoic acid, benzeneacetic acid, 2-carboxy, 2-carboxyphenylacetic acid, 2-carboxybenzeneacetic acid, alpha-carboxy-o-toluic acid, o-carboxymethyl benzoic acid, 2-carboxyphenyl acetic acid, o-toluic acid, alpha-carboxy, toluene-alpha,2-dicarboxylic acid | |
| C1=CC=C(C(=C1)CC(=O)O)C(=O)O | |
| 180.159 | |
| 180.16 | |
| Homophthalic acid |
| ∼180°C (decomposition) | |
| MFCD00004326 | |
| 1872069 | |
| ZHQLTKAVLJKSKR-UHFFFAOYSA-N | |
| 2-(carboxymethyl)benzoic acid | |
| 66643 | |
| 99% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 201-913-9
RTECSNumber : CY1575590
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only