missing translation for 'onlineSavingsMsg'
Learn More

Homophthalic acid, 99%

Catalog No. AAA1429730
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1429730 250 g
AAA1429718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1429730 Supplier Thermo Scientific Chemicals Supplier No. A1429730
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 89-51-0 | C9H8O4 | 180.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89-51-0
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00004326
InChI Key ZHQLTKAVLJKSKR-UHFFFAOYSA-N
Synonym homophthalic acid, 2-carboxymethyl benzoic acid, benzeneacetic acid, 2-carboxy, 2-carboxyphenylacetic acid, 2-carboxybenzeneacetic acid, alpha-carboxy-o-toluic acid, o-carboxymethyl benzoic acid, 2-carboxyphenyl acetic acid, o-toluic acid, alpha-carboxy, toluene-alpha,2-dicarboxylic acid
PubChem CID 66643
IUPAC Name 2-(carboxymethyl)benzoic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)C(=O)O

Specifications

Melting Point ∼180°C (decomposition)
Quantity 250 g
Beilstein 1872069
Formula Weight 180.16
Percent Purity 99%
Chemical Name or Material Homophthalic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-913-9
RTECSNumber CY1575590
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only