missing translation for 'onlineSavingsMsg'
Learn More

Homogentisic Acid 97.0+%, TCI America™

Catalog No. p-7126571
Change view
Click to view available options
Quantity:
1 g
200 mg
5 g
Catalog No. Quantity
D1050200MG 200 mg
D10501G 1 g
D10505G 5 g
3 options

Catalog No. D1050200MG

Supplier: TCI America D1050200MG

Only null left

Chemical Identifiers

CAS 451-13-8
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00004324
InChI Key IGMNYECMUMZDDF-UHFFFAOYSA-N
Synonym homogentisic acid, 2,5-dihydroxyphenylacetic acid, alcapton, homogentisate acid, homogentisinic acid, 2,5-dihydroxyphenyl acetic acid, homogentisate, benzeneacetic acid, 2,5-dihydroxy, 2-2,5-dihydroxyphenyl acetic acid, 2,5-dihydroxy-alpha-toluic acid
PubChem CID 780
ChEBI CHEBI:44747
IUPAC Name 2-(2,5-dihydroxyphenyl)acetic acid
SMILES OC(=O)CC1=CC(O)=CC=C1O

Specifications

Melting Point 147°C
Color White-Yellow
Quantity 200 mg
Formula Weight 168.15
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Homogentisic Acid