missing translation for 'onlineSavingsMsg'
Learn More

Holmium, plasma standard solution, Specpure™, Ho 1000μg/mL, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 035756AE

 View more versions of this product

Catalog No. AA35756AE


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Holmium plasma standard solution
Liquid
MFCD00011049
100 mL
Ho 1000μg/mL
Ho2O3 in 5% HNO3
UN3264
Specpure

Chemical Identifiers

MFCD00011049
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314
Causes severe skin burns and eye damage.

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

EINECSNumber : 231-714-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only