missing translation for 'onlineSavingsMsg'
Learn More

Holmium pieces, 99.9% (metals basis excluding Ta)

Catalog No. p-7048198
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA0014206 5 g
AA0014214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA0014206 Supplier Thermo Scientific Chemicals Supplier No. 00014206
Only null left

CAS: 7440-60-0 | Ho | 164.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged in air-stable poly bottle

Chemical Identifiers

CAS 7440-60-0
Molecular Formula Ho
Molecular Weight (g/mol) 164.93
MDL Number MFCD00011049
InChI Key KJZYNXUDTRRSPN-UHFFFAOYSA-N
Synonym atom, unii-w1xx32sqn1, w1xx32sqn1, foil, holmio, chips, ingot, pieces, powder, acmc-1c9ym
PubChem CID 23988
ChEBI CHEBI:49648
IUPAC Name holmium
SMILES [Ho]

Specifications

Quantity 5 g
Packaging Poly bottle
Form Pieces
Percent Purity 99.9%
Assay (metals basis excluding Ta)
Chemical Name or Material Holmium pieces
EINECSNumber 231-169-0
TSCA Yes
storageNote1
  • Packaged in air-stable poly bottle
Recommended Storage Ambient temperatures

RUO – Research Use Only