missing translation for 'onlineSavingsMsg'
Learn More

Holmium(III) oxide, 99.9%, (Total Rare Earth Oxides), -325 mesh

Catalog No. p-3734785
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC197140100 10 g Glass bottle
AC197140500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC197140100 Supplier Thermo Scientific Chemicals Supplier No. 197140100
Only null left

CAS: 12055-62-8 | Ho2O3 | 377.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis

Chemical Identifiers

CAS 12055-62-8
Molecular Formula Ho2O3
Molecular Weight (g/mol) 377.86
MDL Number MFCD00011053
InChI Key OWCYYNSBGXMRQN-UHFFFAOYSA-N
IUPAC Name diholmium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Ho+3].[Ho+3]

Specifications

Boiling Point 3900.0°C
Melting Point 2330.0°C
Color Beige to Orange-Yellow
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.85% min. (Ho2O3) TREO base
Linear Formula HO2O3
Merck Index 15, 4765
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in inorganic acids
Formula Weight 377.85
Percent Purity 99.9%
Packaging Glass bottle
Chemical Name or Material Holmium(III) oxide
Show More Show Less
EINECSNumber 235-015-3

RUO – Research Use Only