missing translation for 'onlineSavingsMsg'
Learn More

Holmium(III) fluoride, anhydrous, 99.9% (REO)

Catalog No. p-7059620
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4503309 10 g
AA4503318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4503309 Supplier Thermo Scientific Chemicals Supplier No. 04503309
Only null left

CAS: 13760-78-6 | F3Ho | 221.93 g/mol

Holmium(III) fluoride, anhydrous is used to produce the strongest artificially- generated magnetic fields, nuclear control rods, yellow-colored glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13760-78-6
Molecular Formula F3Ho
Molecular Weight (g/mol) 221.93
MDL Number MFCD00016142
InChI Key FDIFPFNHNADKFC-UHFFFAOYSA-K
IUPAC Name holmium(3+) trifluoride
SMILES [F-].[F-].[F-].[Ho+3]

Specifications

Melting Point 1143°C
Physical Form Powder
Quantity 10 g
UN Number UN3288
Solubility Information Insoluble in water.
Formula Weight 221.93
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 7.6 g/mL
Assay (REO)
Chemical Name or Material Holmium (III) fluoride, Anhydrous
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 237-352-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only