CAS RN 13760-78-6
IUPAC Name:
holmium(3+) trifluoride
Synonyms:
holmium(3+) trifluoride
Molecular Weight (g/mol):
221.93
Molecular Formula:
F3Ho
InChi Key:
FDIFPFNHNADKFC-UHFFFAOYSA-K
SMILES:
[F-].[F-].[F-].[Ho+3]
1
–
1
of
1
results
1
Holmium(III) fluoride, anhydrous, 99.9% (REO)
CAS: 13760-78-6 Molecular Formula: F3Ho Molecular Weight (g/mol): 221.93 MDL Number: MFCD00016142 InChI Key: FDIFPFNHNADKFC-UHFFFAOYSA-K IUPAC Name: holmium(3+) trifluoride SMILES: [F-].[F-].[F-].[Ho+3]