missing translation for 'onlineSavingsMsg'
Learn More

Holmium(III) chloride, ultra dry, 99.95% (metals basis)

Catalog No. p-7052311
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4717806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4717806 Supplier Thermo Scientific Chemicals Supplier No. 04717806

CAS: 10138-62-2 | Cl3Ho | 271.28 g/mol

It is mainly used in research. It is used in ceramics, lasers, nuclear industries and in paramagnetic. It is also employed in calibration of optical spectrophotometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon

Chemical Identifiers

CAS 10138-62-2
Molecular Formula Cl3Ho
Molecular Weight (g/mol) 271.28
MDL Number MFCD00011051
InChI Key PYOOBRULIYNHJR-UHFFFAOYSA-K
Synonym holmium chloride, holmium trichloride, holmium chloride hocl3, holmium iii chloride, wln: ho n-o3*3, holmium iii chloride, anhydrous, holmium iii chloride, anhydrous, ?hocl3, holmium iii chloride, anhydrous, powder trace metals basis, 3ar,8ar---4,4,8,8-tetrakis 3,5-diethylphenyl tetrahydro-2,2-dimethyl-6-phenyl-1,3-dioxolo 4,5-e dioxaphosphepin
PubChem CID 24992
IUPAC Name holmium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Ho+3]

Specifications

Melting Point 718°C
Physical Form Beads
Quantity 5 g
Merck Index 14,4727
Solubility Information Soluble in water.
Formula Weight 271.29
Percent Purity 99.95%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 3.70 g/mL
Assay (metals basis)
Chemical Name or Material Holmium(III) chloride, Ultra dry
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-387-1
RTECSNumber MS7350000
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only