CAS RN 10138-62-2
IUPAC Name:
holmium(3+) trichloride
Synonyms:
holmium(III) chloride
Molecular Weight (g/mol):
271.28
Molecular Formula:
Cl3Ho
InChi Key:
PYOOBRULIYNHJR-UHFFFAOYSA-K
SMILES:
[Cl-].[Cl-].[Cl-].[Ho+3]
1
–
1
of
1
results
1
Holmium(III) chloride, ultra dry, 99.95% (metals basis)
CAS: 10138-62-2 Molecular Formula: Cl3Ho Molecular Weight (g/mol): 271.28 MDL Number: MFCD00011051 InChI Key: PYOOBRULIYNHJR-UHFFFAOYSA-K Synonym: holmium chloride,holmium trichloride,holmium chloride hocl3,holmium iii chloride,wln: ho n-o3*3,holmium iii chloride, anhydrous,holmium iii chloride, anhydrous, ?hocl3,holmium iii chloride, anhydrous, powder trace metals basis,3ar,8ar---4,4,8,8-tetrakis 3,5-diethylphenyl tetrahydro-2,2-dimethyl-6-phenyl-1,3-dioxolo 4,5-e dioxaphosphepin PubChem CID: 24992 IUPAC Name: holmium(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Ho+3]