missing translation for 'onlineSavingsMsg'
Learn More

Holmium foil, 0.62mm (0.024 in.) thick, 99.9% (REO), Thermo Scientific Chemicals

Catalog No. p-7048231
Change view
Click to view available options
Quantity:
25 x 25 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA00596FF 25 x 25 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA00596FF Supplier Thermo Scientific Chemicals Supplier No. 000596FF
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon

Chemical Identifiers

CAS 7440-60-0
Molecular Formula Ho
Molecular Weight (g/mol) 164.93
MDL Number MFCD00011049
InChI Key KJZYNXUDTRRSPN-UHFFFAOYSA-N
Synonym atom, unii-w1xx32sqn1, w1xx32sqn1, foil, holmio, chips, ingot, pieces, powder, acmc-1c9ym
PubChem CID 23988
ChEBI CHEBI:49648
IUPAC Name holmium
SMILES [Ho]

Specifications

Quantity 25 x 25 mm
Odor Odorless
Packaging Mylar under Argon
Weight ≈3.4g/25x25mm
Form Foil
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Holmium foil, 0.62mm (0.024 in.) thick
EINECSNumber 231-169-0
TSCA Yes
storageNote1
  • Packaged under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only