missing translation for 'onlineSavingsMsg'
Learn More

Holmium foil, 0.127mm (0.005in) thick, 99.9% (REO)

Catalog No. AA00144FF
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA00144FF 25 x 25 mm
AA00144FI 50 x 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA00144FF Supplier Thermo Scientific Chemicals Supplier No. 000144FF

Form: Approx.0.70g/25 x 25mm | CAS: 7440-60-0 | Ho | 164.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-60-0
Molecular Formula Ho
Molecular Weight (g/mol) 164.93
MDL Number MFCD00011049
InChI Key KJZYNXUDTRRSPN-UHFFFAOYSA-N
Synonym atom, unii-w1xx32sqn1, w1xx32sqn1, foil, holmio, chips, ingot, pieces, powder, acmc-1c9ym
PubChem CID 23988
ChEBI CHEBI:49648
IUPAC Name holmium
SMILES [Ho]

Specifications

Color Silver to Gray
Physical Form Foil
Quantity 25 x 25 mm
Assay Percent Range 99.9% (REO)
Odor Odorless
Appearance Silver gray foil
Chemical Name or Material Holmium
EINECSNumber 231-169-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only