Learn More
Humboldt Hoffman Tubing Clamps
Regulate flow from fully open to closed
Specifications
Quantity 1 Ea.
Type Tubing Clamp
Material Nickel-plated Brass
For Use With (Equipment) Control The Flow in Flexible Tubing
For Use With (Application) Control the flow in flexible tubing
View More Specs
Products 4
Catalog Number Mfr. No. Size Height (English) Height (Metric) Jaw Type Price Quantity & Availability  
Catalog Number Mfr. No. Size Height (English) Height (Metric) Jaw Type Price Quantity & Availability  
05870A
View Documents Promotion Details
Humboldt
H 8730
0.75 in. 0 to 0.62 in. 0 to 16 mm Fixed N/A
Only null left
05871B
View Documents Promotion Details
Humboldt
H 8680
1 in. 0 to 0.75 in. 0 to 19 mm Swivel N/A
Only null left
05870B
View Documents Promotion Details
Humboldt
H 8740
1 in. 0 to 0.75 in. 0 to 19 mm Fixed N/A
Only null left
05871A
View Documents Promotion Details
Humboldt
H 8660
0.75 in. 0 to 0.62 in. 0 to 16 mm Swivel N/A
Only null left
Description
Description
Regulate flow from fully open to closed
  • For flexible tubing
  • Nickel-plated brass
  • Won't rust
  • Lower jaw is either fixed or swivel-mounted for easy insertion of tubing without disconnecting apparatus
Specifications
Specifications
1 Ea.
Nickel-plated Brass
Control the flow in flexible tubing
Tubing Clamp
Control The Flow in Flexible Tubing