Learn More
Humboldt Hoffman Tubing Clamps
Regulate flow from fully open to closed
Supplier: Humboldt H 8740
Description
Regulate flow from fully open to closed
- For flexible tubing
- Nickel-plated brass
- Won't rust
- Lower jaw is either fixed or swivel-mounted for easy insertion of tubing without disconnecting apparatus
Chemical Identifiers
| 1 Ea. | |
| Nickel-plated Brass | |
| Control the flow in flexible tubing |
| Tubing Clamp | |
| Control The Flow in Flexible Tubing |
Specifications
| 1 Ea. | |
| Nickel-plated Brass | |
| Control the flow in flexible tubing | |
| 0 to 0.75 in. | |
| Fixed |
| Tubing Clamp | |
| Control The Flow in Flexible Tubing | |
| 0 to 19 mm | |
| 1 in. |