Learn More
Hoefer™ SUB6 Mini Submarine Combs DFS Item
Accessories for SUB6 Mini Submarine
Supplier:  Hoefer™ SUB6C28
 View more versions of this product
Catalog No. 03-500-703
Only null left
Explore available promotions
Description
Description
  • Ranges from 8, 12 and 16 Wells
  • 1, 1.5 and 2 mm Thickness
  • 2.2, 2.5 and 4.5 mm Well Width
  • Mini. 10, max. 40 μL Sample Volume
Specifications
Specifications
SUB6 Mini Submarine
8
0.2 cm
0.17 in.
0.19 in.
Comb
0.07 in.
40 μL
0.45 cm
0.5 cm
Product Suggestions
Product Suggestions