Learn More
Hoefer™ SUB6 Mini Submarine Combs
Accessories for SUB6 Mini Submarine
Supplier: Hoefer™ SUB6C18
Description
- Ranges from 8, 12 and 16 Wells
- 1, 1.5 and 2 mm Thickness
- 2.2, 2.5 and 4.5 mm Well Width
- Mini. 10, max. 40 μL Sample Volume
Specifications
| SUB6 Mini Submarine | |
| 8 | |
| 0.1 cm | |
| 0.17 in. | |
| 0.19 in. |
| Comb | |
| 0.03 in. | |
| 20 μL | |
| 0.45 cm | |
| 0.5 cm |