Learn More
Hoefer™ Spacers for SE 600 Chroma, SE 600 Standard, or SE 400 Vertical Electrophoresis Units
Accessory parts for SE 600 Chroma SE 600 Standard or SE 400 Vertical Electrophoresis Units
Supplier: Hoefer™ SE61192.75
Description
- Ranges from 0.75 to 3.0 mm Thickness
- 8, 16 and 24 cm in Length
- 1 to 2 cm Width
- 2 Pk.
Specifications
| SE 600 Chroma, SE 600 Standard, SE 400 Vertical Electrophoresis Unit | |
| 2 x 16 cm x 0.75mm | |
| 0.02 in. | |
| 16 cm |
| Spacer | |
| 2/Pk. | |
| 0.07 cm | |
| 2 cm |