missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Combs for SE 600 Series or SE 400 Series Vertical Electrophoresis Units

Catalog No. 361003862
Encompass
Change view
Click to view available options
Comb; 15 wells; 0.75mm thick
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
361003862 Comb; 15 wells; 0.75mm thick
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361003862 Supplier Hoefer™ Supplier No. SE511 15 .75

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems

  • Lightweight
  • Virtually unbreakable
  • Come attached to a standard back which produces maximum well depths of 25 mm, except for the 28-well preparative combs which produce well depths of 15 mm
  • Adjustable comb back (available separately, below) converts 25 mm deep combs to 10 or 15 mm depth

Specifications

For Use With (Equipment) SE 600, SE 400 Series Vertical Electrophoresis System
Type Standard Comb
Material PTFE
No. of Wells 15
Thickness (English) 0.02 in.
Thickness (Metric) 0.07 cm
Volume (Metric) Well 4.3μL/ mm
Width (Metric) 0.57 cm
Depth (English) 0.98 in.
Depth (Metric) 2.5 cm