missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Combs for SE 250, SE 260, or miniVE Vertical Electrophoresis Units

Catalog No. 361003886
Encompass
Change view
Click to view available options
15 wells; 0.75mm thick
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
361003886 15 wells; 0.75mm thick
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361003886 Supplier Hoefer™ Supplier No. SE211A 15 .75

Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems

  • Lightweight
  • Virtually unbreakable

Specifications

For Use With (Equipment) SE 250, SE 260, MiniVE™ Vertical Electrophoresis System
Type Comb
Material PTFE
No. of Wells 15
Thickness (English) 0.02 in.
Thickness (Metric) 0.07 cm
Volume (Metric) Well 2.2μL/ mm
Width (Metric) 0.29 cm
Depth (English) 0.51 in.
Depth (Metric) 1.3 cm