missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Combs for SE 250, SE 260, or miniVE Vertical Electrophoresis Units

Catalog No. 361003885
Encompass
Change view
Click to view available options
No. of Wells:
1 Prep, 1 Reference
10
15
18
5
Thickness (Metric):
0.07 cm
0.075 cm
0.1 cm
0.15 cm
Volume (Metric) Well:
120μL/ mm
13μL/ mm
19.5μL/ mm
2.2μL/ mm
2.9μL/ mm
4.4μL/ mm
4.8μL/ mm
51μL/ mm
68μL/ mm
7.2μL/ mm
9.8μL/ mm
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells Thickness (Metric) Volume (Metric) Well
361003885 15 0.1 cm 2.9μL/ mm
03500169 18 0.1 cm 2.9μL/ mm
03500176 1 Prep, 1 Reference 0.15 cm 120μL/ mm
03500166 15 0.07 cm 2.2μL/ mm
03500174 1 Prep, 1 Reference 0.07 cm 51μL/ mm
03500163 10 0.075 cm 9.8μL/ mm
03500168 15 0.15 cm 4.4μL/ mm
03500165 10 0.15 cm 7.2μL/ mm
03500172 5 0.15 cm 19.5μL/ mm
03500164 10 0.1 cm 4.8μL/ mm
361003881 5 0.1 cm 13μL/ mm
03500175 1 Prep, 1 Reference 0.1 cm 68μL/ mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 361003885 Supplier Hoefer™ Supplier No. SE211A 15 1.0
Only null left

Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems

  • Lightweight
  • Virtually unbreakable

Specifications

For Use With (Equipment) SE 250, SE 260, MiniVE™ Vertical Electrophoresis System
Type Comb
Material PTFE
No. of Wells 15
Thickness (English) 0.03 in.
Thickness (Metric) 0.1 cm
Volume (Metric) Well 2.9μL/ mm
Width (Metric) 0.29 cm
Depth (English) 0.51 in.
Depth (Metric) 1.3 cm