missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ Combs for SE 250, SE 260, or miniVE Vertical Electrophoresis Units
Description
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
- Lightweight
- Virtually unbreakable
Specifications
Specifications
| For Use With (Equipment) | SE 250, SE 260, MiniVE™ Vertical Electrophoresis System |
| Type | Comb |
| Material | PTFE |
| No. of Wells | 18 |
| Thickness (English) | 0.03 in. |
| Thickness (Metric) | 0.1 cm |
| Volume (Metric) Well | 2.9μL/ mm |
| Width (Metric) | 0.29 cm |
| Depth (English) | 0.51 in. |
| Depth (Metric) | 1.3 cm |