Learn More
Hoefer™ Combs for HE 99X Max Submarine Electrophoresis Unit
Catalog No. 03500146
Encompass
Change view
Click to view available options
Type:
Backless Comb
Comb Back
No. of Wells:
10
15
20
30
Thickness (Metric):
0.1 cm
0.15 cm
0.3 cm
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type No. of Wells Thickness (Metric)
03500146 Comb Back - -
03500135 Backless Comb 10 0.15 cm
03500136 Backless Comb 10 0.3 cm
03500143 Backless Comb 30 0.1 cm
03500141 Backless Comb 20 0.15 cm
03500138 Backless Comb 15 0.15 cm
03500139 Backless Comb 15 0.3 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 03500146 Supplier Hoefer™ Supplier No. HFRHE91BK
Requires
Comb back must be ordered separately (see ordering table)
Backless combs for use with Hoefer HE99X Max Submarine Electrophoresis Unit
Combs can be used interchangeably with a single comb back.
Specifications
For Use With (Equipment) HE99X Max Submarine Electrophoresis Unit
Type Comb Back
Includes 2 Screws
Depth (English) Pan 0.19 in.
Depth (Metric) Pan 0.5 cm