missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Hoechst 33342 Ready Flow™ Reagent

Catalog No. R37165
Change view
Click to view available options
Quantity:
120 assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R37165 120 assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R37165 Supplier Invitrogen™ Supplier No. R37165

Hoechst 33342 Ready Flow™ Reagent is a bright, easy-to-use cell-permeant stain for cell cycle analysis on a flow cytometer.

This dye emits blue fluorescence when bound to double-stranded DNA with an emission maximum at 461 nm.

• Stable at room temperature
• Convenient, ready-to-use format—no need to dilute, weigh, or pipette
• Cell-permeable and effective in fixed and live-cell staining for cell cycle

Hoechst 33342 Ready Flow Reagent is a cell-permeable, blue fluorescent DNA stain for cell cycle analysis. Add 2 drops from the dropper bottle directly into 1 x 106 cells, incubate, and analyze.

Hoechst 33342 reagent binds preferentially to adenine-thymine regions of double-stranded DNA. This dye binds into the minor grooves of DNA and exhibits distinct fluorescence emission spectra that are dependent on dye:base pair ratios. Hoechst 33342 Ready Flow Reagent is excited by UV light and has peak emission at 460 nm.

Specifications

Color Blue
Content And Storage 2 dropper bottles, store at room temperature
Excitation Wavelength Range 361/497
Dye Type Cell-Permeant
Quantity 120 assays
Product Line Ready Flow
Product Type Nucleic Acid Stain
Sub Cellular Localization Nucleus
Is DAPI a good live-cell nuclear label?

DAPI is considered a semi-permeant/impermeant nucleic acid stain. Staining of nucleic is dependent upon the cell line in its performance. Some cell lines will label with DAPI, others not at all, and others label inconsistenly. Instead, we recommend using either Hoechst 33342 or Hoechst 33258, which have the same wavelength and binding mode as DAPI (at the A-T minor groove) but are readily cell-permeant.

I want to label the nuclei of live cells and track them over time. Can I use DAPI for this?

We do not recommend doing this. DAPI is considered to be a semi-permeant/impermeant nucleic acid stain. DAPI staining of live cells may be inconsistent. It is best used as a counterstain for fixed samples. Other cell permeable nucleic acid stains, such as Hoechst or the SYTO dyes may affect cellular function.

For mammalian cells, we recommend using the CellLight Nucleus transduction reagents, available in CFP, GFP and RFP. With these reagents, the cells are transduced overnight in a single labeling step and the next day the nuclei will fluoresce. The label may be retained for 3-5 days and should not affect cell function. Cytoplasmic cell tracking dyes such as the CellTracker dyes may also be used.

I would like to label only 100 µL of sample of the same cell density using a Ready Flow product. The manual notes 2 drops per 1 mL (1 X 10E6 cells/mL). May I scale down for smaller volumes?

Yes, you may scale up or down as needed, but we recommend keeping the cell density the same. There are 41 µL per drop, so 2 drops is 82 µL (for the 1 mL of sample). You need to only scale down for this 100 µL volume, which would be 8.2 µL.

What is the concentration of dye/reagent in the Ready Flow products?

The amount of dye or reagent in the Ready Flow products is proprietary. If you need to consider the exact amount of dye or reagent for your experiment, each of the Ready Flow reagents is available as a standalone product.

Can I use the ReadyProbes reagents for flow cytometry?

This is not recommended. The ReadyProbes reagents were developed for imaging applications whereas the Ready Flow reagents were optimized for flow cytometry.