missing translation for 'onlineSavingsMsg'
Learn More

HNRNPF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102794
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102794 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102794 Supplier Thermo Fisher Scientific Supplier No. PA5102794

HNRNPF Polyclonal Antibody

HNRNPF Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Specifications

Antigen HNRNPF
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Hnrnpf
Gene Accession No. P52597, Q794E4, Q9Z2X1
Gene Alias 4833420I20Rik; AA407306; heterogeneous nuclear ribonucleoprotein F; Heterogeneous nuclear ribonucleoprotein F, N-terminally processed; hnRNP F; HnRNP F protein; hnRNP H; Hnrnpf; HNRNPH1; Hnrpf; mcs94 1; mcs94-1; Nucleolin-like protein mcs94-1; OK/SW-cl.23; ribonucleoprotein F; ROF
Gene Symbols Hnrnpf
Host Species Rabbit
Immunogen A synthesized peptide derived from human HNRNPF(Accession P52597), corresponding to amino acid residues V27-S77.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3185, 64200, 98758
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less