missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ hnRNP K Recombinant Rabbit Monoclonal Antibody (23GB4825)

Catalog No. MA553813
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553813 20 μL
MA553812 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553813 Supplier Invitrogen™ Supplier No. MA553813
Only null left

Rabbit Recombinant Monoclonal Antibody

hnRNP K Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene is located in the nucleoplasm and has three repeats of KH domains that binds to RNAs. It is distinct among other hnRNP proteins in its binding preference; it binds tenaciously to poly(C). This protein is also thought to have a role during cell cycle progression. Several alternatively spliced transcript variants have been described for this gene, however, not all of them are fully characterized.

Specifications

Antigen hnRNP K
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4825
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HNRNPK
Gene Accession No. P61978, P61979, P61980
Gene Alias AUKS; CSBP; dC stretch-binding protein; dC-stretch binding protein; FLJ41122; heterogeneous nuclear ribonucleoprotein K; heterogeneous nuclear ribonucleoprotein K S homeolog; hnRNP K; Hnrnpk; hnRNP-K protein; hnrnpk.S; hnrpk; KBBP; NOVA; RP11-575L7.1; transformation upregulated nuclear protein; Transformation up-regulated nuclear protein; TUNP; XELAEV_18010151mg
Gene Symbols HNRNPK
Host Species Rabbit
Immunogen A synthesized peptide derived from human hnRNP K (1-53AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117282, 15387, 3190
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less