missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HN1 Recombinant Rabbit Monoclonal Antibody (23GB6065)

Catalog No. MA552979
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552979 20 μL
MA552978 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552979 Supplier Invitrogen™ Supplier No. MA552979
Only null left

Rabbit Recombinant Monoclonal Antibody

HN1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

HN1 (hematopoietic- and neurologic-expressed sequence 1) is a highly conserved protein that is expressed in developing and regenerating tissues. Overexpression of HN1 has been reported in various tumors like glioma and breast cancer. Hn1 has also been reported as a marker for human ovarian carcinoma and can distinguish epithelial ovarian carcinoma cells from normal ovarian surface epithelial cells. This collective information suggests that HN1 is involved in processes associated with cell proliferation, repair and/or growth.

Specifications

Antigen HN1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB6065
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene JPT1
Gene Accession No. P97825, Q6AXU6, Q9UK76
Gene Alias Androgen-regulated protein 2; ARM2; hematological and neurological expressed 1; hematological and neurological expressed 1 protein; hematological and neurological expressed sequence 1; HN1; HN1A; Jpt1; Jupiter microtubule associated homolog 1; Jupiter microtubule associated homolog 1, N-terminally processed
Gene Symbols JPT1
Host Species Rabbit
Immunogen A synthesized peptide derived from human HN1 (100-154AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15374, 287828, 51155
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less