missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ HMIG Labels
Description
- Made of self-adhesive high-gloss paper
- Available in size: W x H: 4 x 5.875 in.
Specifications
Specifications
| Adhesive Type | Acrylic, Permanent |
| Color | Black, Red, Blue, Yellow, White |
| Finish | Semi-gloss |
| Format | Label |
| Material | Paper (B-235) |
| Product Type | HMIG Label |
| Display Text | Header: HAZARD RATING INDEX, Legend: HAZARDOUS MATERIAL IDENTIFICATION GUIDE |
| Shape | Rectangle |
| Size (English) | 4 x 5.875 in. (W x H) |
| Length (English) | 5.875 in. |
| Show More |